accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 650628-72-1

IUPAC Name: 4-[(pyrrolidin-1-yl)methyl]benzaldehyde
Synonyms: 4-(pyrrolidin-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: MTBYCACQSYAYCC-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results