accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65086-12-6

IUPAC Name: 1,2,3-trimethyl-1H-imidazol-3-ium methyl sulfate
Synonyms: 1,2,3-trimethylimidazol-1-ium methyl sulfate(1-)
Molecular Weight (g/mol): 222.26
Molecular Formula: C7H14N2O4S
InChi Key: OUAUEIYYLHUEPK-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.CN1C=C[N+](C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results