Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trimethylimidazolium methyl sulfate, 95%, Thermo Scientific™

Catalog No. AAH5972422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5972422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5972422 Supplier Thermo Scientific Chemicals Supplier No. H5972422

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65086-12-6
Molecular Formula C7H14N2O4S
Molecular Weight (g/mol) 222.259
MDL Number MFCD03788934
InChI Key OUAUEIYYLHUEPK-UHFFFAOYSA-M
Synonym 1,2,3-trimethylimidazolium methyl sulfate, basionic tm st 99, 1,2,3-trimethylimidazol-1-ium methyl sulfate 1-, basionics™ st 99, 1,2,3-trimethylimidazolium methanesulfonate, 1,2,3-trimethylimidazolium methyl sulfate, basf quality
PubChem CID 11579446
IUPAC Name methyl sulfate;1,2,3-trimethylimidazol-1-ium
SMILES CC1=[N+](C=CN1C)C.COS(=O)(=O)[O-]

Specifications

Melting Point 115°C
Flash Point 196°C (385°F)
Quantity 100 g
Sensitivity Moisture sensitive
Formula Weight 222.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1,2,3-Trimethylimidazolium methyl sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.