accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFFFFFF

CAS RN 651-83-2

IUPAC Name: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
Synonyms: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 233.09
Molecular Formula: C7H2F7N
InChi Key: FJOACTZFMHZHSC-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)