Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%

Catalog No. AAB2432606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2432606 5 g
AAB2432603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2432606 Supplier Thermo Scientific Chemicals Supplier No. B2432606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 651-83-2 | C7H2F7N | 233.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 651-83-2
Molecular Formula C7H2F7N
Molecular Weight (g/mol) 233.089
MDL Number MFCD00091518
InChI Key FJOACTZFMHZHSC-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-trifluoromethyl aniline, 2,3,5,6-tetrafluoro-4-aminobenzotrifluoride, 4-amino-heptafluorotoluene, 4-amino-2,3,5,6-tetrafluorobenzotrifluoride, 4-aminotetrafluorobenzotrifluoride, 4-aminoheptafluorotoluene, 2,3,5,6-tetrafluoro-4-trifluoromethyl phenylamine, benzenamine, 2,3,5,6-tetrafluoro-4-trifluoromethyl, pubchem2817, yu-wasa auxiliary
PubChem CID 616257
IUPAC Name 2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(F)(F)F

Specifications

Boiling Point 185°C to 187°C
Refractive Index 1.43
Quantity 5 g
UN Number UN2810
Formula Weight 233.09
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
Hazard Category H227-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.