accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65104-06-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 298.12
Molecular Formula: C6H22N2O8Ti
InChi Key: PCIGTTWKYUNLEP-UHFFFAOYNA-N
SMILES: N.N.O.O.[Ti].CC(O)C(O)=O.CC(O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)