Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Titanium(Iv) Bis(Ammoniu mLactato)Dihydroxide Solution

Catalog No. 388165500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-816-5500ML 500 mL
38-816-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 38-816-5500ML Supplier Sigma Aldrich Supplier No. 388165500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Titanium(IV) bis(ammoniu mLactato)dihydroxide can be used as:A water-soluble Ti precursor for the synthesis of TiO2/WO3 nanofibers applicable in photocatalysis.A reactant to synthesize a complex with pyrocatechol violet applicable as a colorimetric HF sensing dye.A Ti precursor to synthesize mesoporous silica loaded with titania nanoparticles for photocatalytic applications.

Specifications

CAS 65104-06-5
Density 1.222 g/mL at 25°C
Molecular Formula C6H18N2O8Ti
Refractive Index n20/D 1.438
Linear Formula [CH3CH(O-)CO2NH4]2Ti(OH)2
MDL Number MFCD00072736
Quantity 500 mL
Synonym Dihydroxybis(ammoniu mLactato)titanium solution
Molecular Weight (g/mol) 294.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.