CAS RN 6512-32-9
IUPAC Name:
methyl 2-(3,5-dimethoxyphenyl)acetate
Synonyms:
methyl 2-(3,5-dimethoxyphenyl)acetate
Molecular Weight (g/mol):
210.23
Molecular Formula:
C11H14O4
InChi Key:
CLZWNNSTRIEAMH-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC(OC)=CC(OC)=C1
1
–
1
of
1
results
1
Methyl (3,5-Dimethoxyphenyl)acetate 98.0+%, TCI America™
CAS: 6512-32-9 Molecular Formula: C11H14O4 Molecular Weight (g/mol): 210.229 MDL Number: MFCD00089971 InChI Key: CLZWNNSTRIEAMH-UHFFFAOYSA-N Synonym: (3,5-Dimethoxyphenyl)acetic Acid Methyl Ester PubChem CID: 592191 IUPAC Name: methyl 2-(3,5-dimethoxyphenyl)acetate SMILES: COC1=CC(=CC(=C1)CC(=O)OC)OC