Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (3,5-Dimethoxyphenyl)acetate 98.0+%, TCI America™
SDP

Catalog No. M29335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29335G 5 g
M29331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29335G Supplier TCI America Supplier No. M29335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6512-32-9
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00089971
InChI Key CLZWNNSTRIEAMH-UHFFFAOYSA-N
Synonym (3,5-Dimethoxyphenyl)acetic Acid Methyl Ester
PubChem CID 592191
IUPAC Name methyl 2-(3,5-dimethoxyphenyl)acetate
SMILES COC1=CC(=CC(=C1)CC(=O)OC)OC

Specifications

Boiling Point 94°C
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (3,5-Dimethoxyphenyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.