accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65131-09-1

IUPAC Name: 3-(benzenesulfonyl)-7-methoxy-1,3-dihydro-2-benzofuran-1-one
Synonyms: 3-(benzenesulfonyl)-7-methoxy-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 304.32
Molecular Formula: C15H12O5S
InChi Key: BNOAFLKWYXXZHU-UHFFFAOYNA-N
SMILES: COC1=CC=CC2=C1C(=O)OC2S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results