accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COONH2NH

CAS RN 6519-67-1

IUPAC Name: ethyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Synonyms: ethyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Molecular Weight (g/mol): 267.73
Molecular Formula: C13H16ClN2O2
InChi Key: KQMBXHHYHOTYOO-UHFFFAOYNA-N
SMILES: [Cl].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results