Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Tryptophan Ethyl Ester Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. T07545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07545G Supplier TCI America Supplier No. T07545G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6519-67-1
Molecular Formula C13H16ClN2O2
Molecular Weight (g/mol) 267.73
MDL Number MFCD00067553
InChI Key KQMBXHHYHOTYOO-UHFFFAOYNA-N
Synonym (+/-)-2-Amino-3-(3-indolyl)propionic Acid Ethyl Ester Hydrochloride, H-DL-Trp-OEt.HCl
PubChem CID 11173230
IUPAC Name ethyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
SMILES [Cl].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 268.74
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Tryptophan Ethyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.