accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNClNOO

CAS RN 65224-66-0

IUPAC Name: 2-chloro-6-methyl-5-nitro-1,4-dihydropyrimidin-4-one
Synonyms: 2-chloro-6-methyl-5-nitro-1H-pyrimidin-4-one
Molecular Weight (g/mol): 189.56
Molecular Formula: C5H4ClN3O3
InChi Key: GNXOLOBSWUZOEF-UHFFFAOYSA-N
SMILES: CC1=C(C(=O)N=C(Cl)N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results