accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6526-72-3

IUPAC Name: 1-nitro-2-(propan-2-yl)benzene
Synonyms: 1-nitro-2-isopropylbenzene
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: BSMKYQUHXQAVKG-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results