Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrocumene 97.0+%, TCI America™
SDP

Catalog No. N053225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N053225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N053225G Supplier TCI America Supplier No. N053225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6526-72-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00039739
InChI Key BSMKYQUHXQAVKG-UHFFFAOYSA-N
Synonym 2-Nitroisopropylbenzene
PubChem CID 81013
IUPAC Name 1-nitro-2-(propan-2-yl)benzene
SMILES CC(C)C1=CC=CC=C1[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 121°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitrocumene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.