accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBr

CAS RN 6529-53-9

IUPAC Name: 1-(2-bromoethyl)-4-chlorobenzene
Synonyms: 1-(2-bromoethyl)-4-chlorobenzene
Molecular Weight (g/mol): 219.51
Molecular Formula: C8H8BrCl
InChi Key: YAFMYKFAUNCQPU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CCBr)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results