accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65291-30-7

O(R)O

CAS RN 65291-30-7

IUPAC Name: (2R)-2-[(triphenylmethoxy)methyl]oxirane
Synonyms: (2R)-2-[(triphenylmethoxy)methyl]oxirane
Molecular Weight (g/mol): 316.4
Molecular Formula: C22H20O2
InChi Key: XFSXUCMYFWZRAF-OAQYLSRUSA-N
SMILES: C(OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[C@H]1CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)