Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Glycidyl Trityl Ether 98.0+%, TCI America™
SDP

Catalog No. G02845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02845G 5 g
G028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02845G Supplier TCI America Supplier No. G02845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65291-30-7
Molecular Formula C22H20O2
Molecular Weight (g/mol) 316.4
MDL Number MFCD00273368
InChI Key XFSXUCMYFWZRAF-OAQYLSRUSA-N
Synonym (R)-Glycidyl Triphenylmethyl Ether, (R)-2-(Triphenylmethoxymethyl)oxirane
PubChem CID 2734442
IUPAC Name (2R)-2-(trityloxymethyl)oxirane
SMILES C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Formula Weight 316.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Glycidyl Trityl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.