CAS RN 6537-46-8
IUPAC Name:
10H-chromeno[3,2-b]pyridin-10-one
Synonyms:
chromeno[3,2-b]pyridin-10-one
Molecular Weight (g/mol):
197.19
Molecular Formula:
C12H7NO2
InChi Key:
YAUODDQCNOBEDJ-UHFFFAOYSA-N
SMILES:
O=C1C2=CC=CC=C2OC2=CC=CN=C12
1
–
2
of
2
results
1
5H-Chromeno[2,3-b]pyridin-5-one, 97%, Thermo Scientific™
CAS: 6537-46-8 Molecular Formula: C12H7NO2 MDL Number: MFCD00052253
2
Promotions Available
4-Azaxanthone, 97%
4-Azaxanthone, 97%
CAS: 6537-46-8 Molecular Formula: C12H7NO2 MDL Number: MFCD00052253