Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5H-Chromeno[2,3-b]pyridin-5-one, 97%, Thermo Scientific™

Catalog No. BTB03140DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB03140DE 5 g
BTB03140DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB03140DE Supplier Thermo Scientific Chemicals Supplier No. BTB03140DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6537-46-8
Molecular Formula C12H7NO2
MDL Number MFCD00052253

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 197.19
Percent Purity 97%
Chemical Name or Material 5H-Chromeno[2, 3-b]pyridin-5-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.