accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOCH3

CAS RN 6540-98-3

IUPAC Name: 6-hydroxydecan-5-one
Synonyms: 6-hydroxydecan-5-one
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: ORLDHCIQVZTBQQ-UHFFFAOYNA-N
SMILES: CCCCC(O)C(=O)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results