accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3(E)(E)

CAS RN 65405-80-3

IUPAC Name: (3E)-hex-3-en-1-yl (2E)-but-2-enoate
Synonyms: (3E)-hex-3-en-1-yl (2E)-but-2-enoate
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: KITGYVIOYOCIIE-PTFSRLPTSA-N
SMILES: CC\C=C\CCOC(=O)\C=C\C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results