Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-yl Crotonate 95.0+%, TCI America™
SDP

Catalog No. C091925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C091925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C091925ML Supplier TCI America Supplier No. C091925ML

Chemical Identifiers

CAS 65405-80-3
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00059903
InChI Key KITGYVIOYOCIIE-PTFSRLPTSA-N
Synonym Crotonic Acid cis-3-Hexen-1-yl Ester
PubChem CID 124155754
IUPAC Name (3E)-hex-3-en-1-yl (2E)-but-2-enoate
SMILES CC\C=C\CCOC(=O)\C=C\C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 168.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl Crotonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.