accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CCH3OPOOOH3C

CAS RN 654058-04-5

IUPAC Name: 1,3-dimethyl-1H-imidazol-3-ium dimethyl phosphate
Synonyms: 1,3-dimethylimidazolium dimethyl phosphate
Molecular Weight (g/mol): 222.18
Molecular Formula: C7H15N2O4P
InChi Key: GSGLHYXFTXGIAQ-UHFFFAOYSA-M
SMILES: COP([O-])(=O)OC.CN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)