Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-3,3,3-Trifluoro-1,2-Epoxypropane

Catalog No. 665797250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-579-7250MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 66-579-7250MG Supplier Sigma Aldrich Supplier No. 665797250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Precursor to a trifluoromethyl chiral auxiliary which promotes a highly diastereoselective Simmons-Smith cyclopropanation.

Specifications

CAS 654058-04-5
Melting Point 98°C to 102°C
Molecular Formula C10H13BO4
Refractive Index n20/D <1.300
Linear Formula (CH3)CHOC6H5(CHO)B(OH)2
MDL Number MFCD08705272
Quantity 1 g
Synonym (S)-(-)-2-(Trifluoromethyl)oxirane
Molecular Weight (g/mol) 208.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.