accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNOHNOOOH

CAS RN 65420-40-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[(9H-xanthen-9-yl)carbamoyl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[(9H-xanthen-9-yl)carbamoyl]propanoic acid
Molecular Weight (g/mol): 412.44
Molecular Formula: C22H24N2O6
InChi Key: YMGDQLXBNMRJMR-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)