accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHNO

CAS RN 65435-04-3

IUPAC Name: 5-(2-chloroacetyl)-2,3-dihydro-1H-indol-2-one
Synonyms: 5-(2-chloroacetyl)-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 209.63
Molecular Formula: C10H8ClNO2
InChi Key: WXJWBEAGVWVEDM-UHFFFAOYSA-N
SMILES: ClCC(=O)C1=CC=C2NC(=O)CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results