Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Chloroacetyl)oxindole 98.0+%, TCI America™
SDP

Catalog No. C27981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27981G Supplier TCI America Supplier No. C27981G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65435-04-3
Molecular Formula C10H8ClNO2
Molecular Weight (g/mol) 209.629
MDL Number MFCD04115734
InChI Key WXJWBEAGVWVEDM-UHFFFAOYSA-N
Synonym 5-(Chloroacetyl)-2-indolinone
PubChem CID 2060576
IUPAC Name 5-(2-chloroacetyl)-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)C(=O)CCl)NC1=O

Specifications

Melting Point 240°C
Quantity 1 g
Formula Weight 209.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Chloroacetyl)oxindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.