CAS RN 654670-89-0
CAS RN 654670-89-0
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(4-methylphenyl)diphenylmethyl]amino}propanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-{[(4-methylphenyl)diphenylmethyl]amino}propanoic acid
Molecular Weight (g/mol):
582.7
Molecular Formula:
C38H34N2O4
InChi Key:
WDZDBCVDBMWMAM-DHUJRADRSA-N
SMILES:
CC1=CC=C(C=C1)C(NC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Dap Mtt -OH, Advanced ChemTech
CAS: 654670-89-0 Molecular Formula: C38H34N2O4 Molecular Weight (g/mol): 582.70 MDL Number: MFCD02094097 InChI Key: WDZDBCVDBMWMAM-DHUJRADRSA-N IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(4-methylphenyl)diphenylmethyl]amino}propanoic acid SMILES: CC1=CC=C(C=C1)C(NC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1