CAS RN 654671-77-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sitagliptin Phosphate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
An anti-diabetic (antihyperglycemic) drug of the dipeptidyl peptidase-4 (DPP-4) class. Inhibition of the enzyme DPP-4 is thought to increase Glucagon-like peptide-1 (GLP-1) which inhibits glucagon release, stimulates insulin release and lowers blood glucose.