accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6547-53-1

IUPAC Name: 2-[4-(benzyloxy)phenyl]acetic acid
Synonyms: [4-(benzyloxy)phenyl]acetic acid
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: XJHGAJLIKDAOPE-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)