Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyphenylacetic acid, 99%, Thermo Scientific™

Catalog No. AAH5090214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5090214 25 g
AAH5090206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5090214 Supplier Thermo Scientific Chemicals Supplier No. H5090214

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6547-53-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00017540
InChI Key XJHGAJLIKDAOPE-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
PubChem CID 81033
IUPAC Name 2-(4-phenylmethoxyphenyl)acetic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O

Specifications

Melting Point 119°C to 121°C
Quantity 25 g
Formula Weight 242.28
Percent Purity 99%
Chemical Name or Material 4-Benzyloxyphenylacetic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-463-9
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.