accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65490-24-6

IUPAC Name: 7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide hydrochloride
Synonyms: 7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide hydrochloride
Molecular Weight (g/mol): 497.33
Molecular Formula: C22H22Cl2N2O7
InChi Key: ISGAAFMBTIWTEU-UHFFFAOYNA-N
SMILES: Cl.CN(C)C1C2CC3=C(C(O)=C4C(O)=CC=C(Cl)C4=C3C)C(=O)C2(O)C(O)=C(C(N)=O)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)