Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anhydrochlortetracycline hydrochloride, can be used as secondary standard, Thermo Scientific™

Catalog No. AC268205000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC268205000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268205000 Supplier Thermo Scientific Chemicals Supplier No. 268205000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65490-24-6
Molecular Formula C22H22Cl2N2O7
Molecular Weight (g/mol) 497.325
MDL Number MFCD00143616
InChI Key ISGAAFMBTIWTEU-DTTSPEASSA-N
Synonym anhydrochlortetracycline hydrochloride, anhydro chlortetracycline hydrochloride, anhydro chlortetracycline, anhydroaureomycin hydrochloride, anhydrochlortetracyline hydrochloride, 2z,4s,4as,12as-2-amino hydroxy methylidene-7-chloro-4-dimethylamino-10,11,12a-trihydroxy-6-methyl-4a,5-dihydro-4h-tetracene-1,3,12-trione hydrochloride, 4s,12as,4as-4-dimethylamino-7-chloro-3,10,11,12a-tetrahydroxy-6-methyl-1,1 2-dioxo-4,5,12a,4a-tetrahydronaphthacene-2-carboxamide, chloride, 4s,4as,12ar-7-chloro-4-dimethylamino-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide-hydrogen chloride 1/1, 4s,4as,12as-7-chloro-4-dimethylamino-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide hydrochloride, 4s,4as,12as-7-chloro-4-dimethylamino-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4h-tetracene-2-carboxamide hydrochloride
PubChem CID 54710411
IUPAC Name (4S,4aS,12aR)-7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C14)Cl)O)O)O)O)C(=O)N)N(C)C.Cl

Specifications

Color Brown-Red to Orange
Infrared Spectrum Authentic
Quantity 500 mg
Formula Weight 497.33
Percent Purity ≥97.0% (on anhydrous substance), ≥80 wt% (as is, calculated)(HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Anhydrochlortetracycline hydrochloride
Hazard Category LIGHT SENSITIVE
Hazard Statement MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.