accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSF

CAS RN 655-20-9

IUPAC Name: 1-fluoro-2-(methylsulfanyl)benzene
Synonyms: 1-fluoro-2-(methylsulfanyl)benzene
Molecular Weight (g/mol): 142.19
Molecular Formula: C7H7FS
InChi Key: AYTSELVZWGHUEE-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results