Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorothioanisole 98.0+%, TCI America™
SDP

Catalog No. F05655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05655G 5 g
F056525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05655G Supplier TCI America Supplier No. F05655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 655-20-9
Molecular Formula C7H7FS
Molecular Weight (g/mol) 142.19
MDL Number MFCD03093762
InChI Key AYTSELVZWGHUEE-UHFFFAOYSA-N
Synonym 2-Fluorophenyl Methyl Sulfide, 1-Fluoro-2-(methylthio)benzene
PubChem CID 2774762
IUPAC Name 1-fluoro-2-(methylsulfanyl)benzene
SMILES CSC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 142.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorothioanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.