CAS RN 65505-17-1
IUPAC Name:
2-methyl-3-(methyldisulfanyl)furan
Synonyms:
2-methyl-3-(methyldisulfanyl)furan
Molecular Weight (g/mol):
160.25
Molecular Formula:
C6H8OS2
InChi Key:
SRUTWBWLFKSTIS-UHFFFAOYSA-N
SMILES:
CSSC1=C(C)OC=C1
1
–
1
of
1
results
1
2-Methyl-3-(methyldithio)furan 98.0+%, TCI America™
CAS: 65505-17-1 Molecular Formula: C6H8OS2 Molecular Weight (g/mol): 160.249 MDL Number: MFCD00038643 InChI Key: SRUTWBWLFKSTIS-UHFFFAOYSA-N Synonym: Methyl 2-Methyl-3-furyl Disulfide PubChem CID: 47649 IUPAC Name: 2-methyl-3-(methyldisulfanyl)furan SMILES: CC1=C(C=CO1)SSC