Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-(methyldithio)furan 98.0+%, TCI America™
SDP

Catalog No. M18375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18375G 5 g
M18371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18375G Supplier TCI America Supplier No. M18375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65505-17-1
Molecular Formula C6H8OS2
Molecular Weight (g/mol) 160.249
MDL Number MFCD00038643
InChI Key SRUTWBWLFKSTIS-UHFFFAOYSA-N
Synonym Methyl 2-Methyl-3-furyl Disulfide
PubChem CID 47649
IUPAC Name 2-methyl-3-(methyldisulfanyl)furan
SMILES CC1=C(C=CO1)SSC

Specifications

Color Yellow
Boiling Point 37°C
Quantity 5 g
UN Number 2810
Formula Weight 160.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-(methyldithio)furan
RTECSNumber JO1975000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.