accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NBrCl

CAS RN 65550-77-8

IUPAC Name: 2-bromo-5-chloro-3-methylpyridine
Synonyms: 2-bromo-5-chloro-3-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: XEHWMXAUPXFOHZ-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CN=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results