Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-chloro-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B51335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51335G 5 g
B513325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51335G Supplier TCI America Supplier No. B51335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 65550-77-8 | C6H5BrClN | 206.47 g/mol

Chemical Identifiers

CAS 65550-77-8
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.47
MDL Number MFCD03095062
InChI Key XEHWMXAUPXFOHZ-UHFFFAOYSA-N
Synonym 2-Bromo-5-chloro-3-picoline
PubChem CID 12630985
IUPAC Name 2-bromo-5-chloro-3-methylpyridine
SMILES CC1=CC(Cl)=CN=C1Br

Specifications

Melting Point 39°C
Color White-Yellow
Quantity 5 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-5-chloro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.