accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6563-13-9

IUPAC Name: 6-methoxyquinolin-1-ium-1-olate
Synonyms: 6-methoxyquinolin-1-ium-1-olate
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: BWEGRKPOJXNZSK-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(C=CC=[N+]2[O-])=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results