Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxyquinoline N-Oxide 98.0+%, TCI America™
SDP

Catalog No. M29995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29995G 5 g
M29991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29995G Supplier TCI America Supplier No. M29995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6563-13-9 | C10H9NO2 | 175.187 g/mol

Chemical Identifiers

CAS 6563-13-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00009771
InChI Key BWEGRKPOJXNZSK-UHFFFAOYSA-N
PubChem CID 253821
IUPAC Name 6-methoxy-1-oxidoquinolin-1-ium
SMILES COC1=CC2=C(C=C1)[N+](=CC=C2)[O-]

Specifications

Melting Point 110°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxyquinoline N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.