accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNNH3CCH3OOH

CAS RN 65671-54-7

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-5-(dimethylamino)pentanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid
Molecular Weight (g/mol): 260.33
Molecular Formula: C12H24N2O4
InChi Key: HBCLYVIARHJTTL-UHFFFAOYNA-N
SMILES: CN(C)CCCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results