Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid, Thermo Scientific™

Catalog No. MO08534DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO08534DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO08534DA Supplier Thermo Scientific Chemicals Supplier No. MO08534DA
Add to Cart

Chemical Identifiers

CAS 65671-54-7
Molecular Formula C12H24N2O4
Molecular Weight (g/mol) 260.334
InChI Key HBCLYVIARHJTTL-VIFPVBQESA-N
Synonym s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, s-2-boc-amino-5-dimethylamino pentanoic acid, s-2-tert-butoxycarbonyl amino-5-dimethylami, n∼2∼-tert-butoxycarbonyl-n∼5∼,n∼5∼-dimethyl-l-ornithine, s-2-tert-butoxycarbonylamino-5-dimethylamino pentanoic acid, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5,n5-dimethyl, 2s-5-dimethylamino-2-tert-butoxy carbonylamino pentanoic acid
PubChem CID 16105709
IUPAC Name (2S)-5-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)O

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 260.33
Percent Purity 97%
Chemical Name or Material (S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.