accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNH2(S)H2NHClHCl

CAS RN 657-26-1

IUPAC Name: (2S)-2,6-diaminohexanoic acid dihydrochloride
Synonyms: lysine dihydrochloride
Molecular Weight (g/mol): 219.11
Molecular Formula: C6H16Cl2N2O2
InChi Key: JBBURJFZIMRPCZ-XRIGFGBMSA-N
SMILES: Cl.Cl.NCCCC[C@H](N)C(O)=O
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)