Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Lysine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. L013125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L013125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L013125G Supplier TCI America Supplier No. L013125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-(+)-Lysine dihydrochloride, a laboratory reagent with ≥98.0% purity.

Chemical Identifiers

CAS 657-26-1
Molecular Formula C6H16Cl2N2O2
Molecular Weight (g/mol) 219.11
MDL Number MFCD00066389
InChI Key JBBURJFZIMRPCZ-XRIGFGBMSA-N
Synonym l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
PubChem CID 69567
IUPAC Name (2S)-2,6-diaminohexanoic acid dihydrochloride
SMILES Cl.Cl.NCCCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 219.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Lysine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.