accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOO

CAS RN 65709-23-1

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)-2-oxoacetaldehyde
Synonyms: 2-(2H-1,3-benzodioxol-5-yl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 178.14
Molecular Formula: C9H6O4
InChi Key: OUCBKIWZWGSDJL-UHFFFAOYSA-N
SMILES: O=CC(=O)C1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results