accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNNH

CAS RN 65709-27-5

IUPAC Name: 2-(2-methoxyphenyl)piperazine
Synonyms: 2-(2-methoxyphenyl)piperazine
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: XYMDIQVPOIVJPF-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1C1CNCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results