Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methoxyphenyl)piperazine monohydrate, 95%

Catalog No. AAH26408MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26408MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26408MD Supplier Thermo Scientific Chemicals Supplier No. H26408MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 65709-27-5 | C11H16N2O | 192.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65709-27-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD06660448
InChI Key XYMDIQVPOIVJPF-UHFFFAOYSA-N
Synonym 2-2-methoxyphenyl piperazine, 2-2-methoxyphenyl-piperazine, 2-2-methoxy-phenyl-piperazine, 2-methoxyphenyl piperazine, acmc-1bd94, 1-methoxy-2-piperazin-2-ylbenzene
PubChem CID 16740792
IUPAC Name 2-(2-methoxyphenyl)piperazine
SMILES COC1=CC=CC=C1C2CNCCN2

Specifications

Melting Point 77°C to 83°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 210.28 (192.26 Anhydrous)
Percent Purity 95%
Chemical Name or Material 2-(2-Methoxyphenyl)piperazine monohydrate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.