accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClN

CAS RN 6575-00-4

IUPAC Name: 3,5-dichlorobenzonitrile
Synonyms: 3,5-dichlorobenzonitrile
Molecular Weight (g/mol): 172.01
Molecular Formula: C7H3Cl2N
InChi Key: PUJSUOGJGIECFQ-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results